Itaconic acid, 99+%
Itaconic acid, 99+%
Itaconic acid, 99+%
Thermo Scientific Chemicals
Itaconic acid, 99+%
CAS: 97-65-4 | C5H6O4 | 130.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA122812500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialItaconic acid
CAS97-65-4
Health Hazard 1GHS Signal Word: Warning
Melting Point165.0°C to 168.0°C
ColorBeige to White