Isonicotinic acid hydrazide, 99%
Isonicotinic acid hydrazide, 99%
Isonicotinic acid hydrazide, 99%
Isonicotinic acid hydrazide, 99%
Thermo Scientific Chemicals
Isonicotinic acid hydrazide, 99%
CAS: 54-85-3 | C6H7N3O | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic bottle
Catalog number FSA122605000
Price (MYR)
1,108.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,108.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsonicotinic acid hydrazide
Name Note99%
CAS54-85-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement