Isobutyronitrile, 99%
Isobutyronitrile, 99%
Isobutyronitrile, 99%
Isobutyronitrile, 99%
Isobutyronitrile, 99%
CAS: 78-82-0 | C4H7N | 69.11 g/mol
Have Questions?
Catalog number FSA122530010
Price (MYR)
602.00
BOT
Add to cart
Request bulk or custom format
Price (MYR)
602.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsobutyronitrile
Name Note99%
CAS78-82-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Fatal if inhaled.
Highly flammable liquid and vapor.