4-Iodophenol, 99%
4-Iodophenol, 99%
4-Iodophenol, 99%
Thermo Scientific Chemicals
4-Iodophenol, 99%
CAS: 540-38-5 | C6H5IO | 220.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA122390250
Price (MYR)
554.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
554.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Iodophenol
CAS540-38-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.