Iodobenzene, 98%
Iodobenzene, 98%
Iodobenzene, 98%
Thermo Scientific Chemicals
Iodobenzene, 98%
CAS: 591-50-4 | C6H5I | 204.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA122335000
Price (MYR)
2,284.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,284.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIodobenzene
CAS591-50-4
Health Hazard 1GHS Signal Word: Warning
Melting Point-29.0°C
ColorYellow