Inosine, 99%
Inosine, 99%
Inosine, 99%
Thermo Scientific Chemicals
Inosine, 99%
CAS: 58-63-9 | C10H12N4O5 | 268.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA122252500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialInosine
CAS58-63-9
Absorbance1.65 to 1.77 (A250/A260), 0.22 to 0.26 (A280/A260)
Melting Point212°C to 213°C