1H-Indole-3-acetic acid, 98%
1H-Indole-3-acetic acid, 98%
1H-Indole-3-acetic acid, 98%
1H-Indole-3-acetic acid, 98%
Thermo Scientific Chemicals
1H-Indole-3-acetic acid, 98%
1H-Indole-3-acetic acid, CAS # 87-51-4, also known as indole-3-acetic acid, is a monocarboxylic acid mainly used as a plant growth hormone and is the most common naturally occurring plant hormone of the auxin family. | CAS: 87-51-4 | C10H9NO2 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA122161000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1H-Indole-3-acetic acid
CAS87-51-4
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
General Information
• 1H-Indole-3-acetic acid is a plant growth hormone that belongs to the auxin class. It is also considered to be an inducer of plant cell elongation and division that can cause uncontrolled growth
Applications
• This compound is a monocarboxylic acid; an acetic acid in which one of the methyl hydrogens has been replaced by a 1H-indol-3-yl group.
• Beyond its role as a plant hormone, this compound regulates tissue differentiation, apical dominance, and responses to light, gravity, and pathogens
• It can be used as a signaling molecule necessary for development of plant organs and coordination of growth
RUO – Research Use Only
General References:
  1. Fu, S.; Wei, J.; Chen, H.; Liu, Y.; Lu, H.; Chou, J. Indole-3-acetic acid: A widespread physiological code in interactions of fungi with other organisms. Plant Signal Behav. 2015, 10, (8), e1048052.
  2. Cheryl L. Patten; Bernard R. Glick. Bacterial biosynthesis of indole-3-acetic acid. Can. J. Microbiol. 1996, 42, (3), 207-220.
  3. Rigsud, J.; Puppo, A. Indole-3-acetic Acid Catabolism by Soybean Bacteroids. Microbiology. 1975, 88, 223-228.