Iminodibenzyl, 97%
Iminodibenzyl, 97%
Iminodibenzyl, 97%
Thermo Scientific Chemicals
Iminodibenzyl, 97%
CAS: 494-19-9 | C14H13N | 195.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA122071000
Price (MYR)
505.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
505.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIminodibenzyl
Name Note97%
CAS494-19-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: