2-Imidazolidinethione, 98%
2-Imidazolidinethione, 98%
2-Imidazolidinethione, 98%
2-Imidazolidinethione, 98%
Thermo Scientific Chemicals
2-Imidazolidinethione, 98%
CAS: 96-45-7 | C3H6N2S | 102.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA122032500
Price (MYR)
574.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
574.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Imidazolidinethione
Name Note98%
CAS96-45-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger