3-Hydroxy-2-naphthoic acid, 98%
3-Hydroxy-2-naphthoic acid, 98%
3-Hydroxy-2-naphthoic acid, 98%
3-Hydroxy-2-naphthoic acid, 98%
Thermo Scientific Chemicals
3-Hydroxy-2-naphthoic acid, 98%
CAS: 92-70-6 | C11H8O3 | 188.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA121611000
Price (MYR)
201.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
201.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Hydroxy-2-naphthoic acid
Name Note98%
CAS92-70-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning