4-Hydroxy-4-methyl-2-pentanone, 99%
4-Hydroxy-4-methyl-2-pentanone, 99%
4-Hydroxy-4-methyl-2-pentanone, 99%
4-Hydroxy-4-methyl-2-pentanone, 99%
Thermo Scientific Chemicals
4-Hydroxy-4-methyl-2-pentanone, 99%
CAS: 123-42-2 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number FSA121482500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Hydroxy-4-methyl-2-pentanone
CAS123-42-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
If eye