Diethylene glycol, 99%, extra pure
Diethylene glycol, 99%, extra pure
Diethylene glycol, 99%, extra pure
Diethylene glycol, 99%, extra pure
Thermo Scientific Chemicals
Diethylene glycol, 99%, extra pure
CAS: 111-46-6 | C4H10O3 | 106.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic drum
Catalog number FSA121160100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethylene glycol
CAS111-46-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0