Clear Glass Graduated Reagent Bottles with Screw Caps and Drip-Free Pouring Rings
Clear Glass Graduated Reagent Bottles with Screw Caps and Drip-Free Pouring Rings
Actual product may vary
Clear Glass Graduated Reagent Bottles with Screw Caps and Drip-Free Pouring Rings
Clear Glass Graduated Reagent Bottles with Screw Caps and Drip-Free Pouring Rings
Clear Glass Graduated Reagent Bottles with Screw Caps and Drip-Free Pouring Rings
Thermo Scientific Chemicals
Clear Glass Graduated Reagent Bottles with Screw Caps and Drip-Free Pouring Rings
These reusable Reagent Bottles are strong and chemical resistant.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FSA121080250250 mL
ACR121080250250 mL
Catalog number FSA121080250
Price (MYR)
559.00
BOT
Add to cart
Capacity (Metric):
250 mL
Price (MYR)
559.00
BOT
Add to cart
  • In conformity to DIN ISO 4796-1 Standard
  • Printed in white color
  • These bottles are mechanically strong and chemical resistant
  • Provided with a pouring ring for drip-free operation
  • The screw caps and pouring ring are made of PP
  • The bottles screw caps and pouring rings can be sterilized and are autoclavable
  • 100 mL to 1000 mL bottles have uniform GL 45 thread
Specifications
Capacity250 mL
Capacity (Metric)250 mL
Capacity (English)8.5 oz.
ColorTransparent
Diameter (English)2.8 ±0.06 in.
For Use With (Application)Storage
IncludesDrip-Free Pouring Rings
MaterialBorosilicate Glass (3.3)
Quantity10 Pk.
SterilitySterile
Certifications/ComplianceDIN ISO 4796-1
Closure IncludedYes
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
MouthThreaded
ShapeRound
Height (Metric)13.80 cm
Product TypeReagent Bottle
For Use With (Equipment)Chemicals
Height (English)5.43 in.
Chemical LabelCustom Write-on
Closure LinerUnlined
Closure MaterialPolypropylene
Closure SizeGL -45
Diameter (Metric)70 ±1.5 mm
Closure TypeScrew Cap
Unit SizeBOT