4-Hydroxybenzoic Acid, 99+%
4-Hydroxybenzoic Acid, 99+%
4-Hydroxybenzoic Acid, 99+%
4-Hydroxybenzoic Acid, 99+%
Thermo Scientific Chemicals
4-Hydroxybenzoic Acid, 99+%
CAS: 99-96-7 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA120995000
Price (MYR)
419.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
419.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Hydroxybenzoic acid
Name Note99%
CAS99-96-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger