3-Hydroxybenzoic acid, 99%
3-Hydroxybenzoic acid, 99%
3-Hydroxybenzoic acid, 99%
Thermo Scientific Chemicals
3-Hydroxybenzoic acid, 99%
CAS: 99-06-9 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number FSA120981000
Price (MYR)
311.00
Bottle
Quantity:
100 g
Request bulk or custom format
Price (MYR)
311.00
Bottle
Specifications
Chemical Name or Material3-Hydroxybenzoic acid
Name Note99%
CAS99-06-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning