1,6-Hexanedithiol, 97%
1,6-Hexanedithiol, 97%
1,6-Hexanedithiol, 97%
1,6-Hexanedithiol, 97%
Thermo Scientific Chemicals
1,6-Hexanedithiol, 97%
CAS: 1191-43-1 | C6H14S2 | 150.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA120670250
Price (MYR)
1,062.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,062.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 6-Hexanedithiol
Name Note97%
CAS1191-43-1
CAS Min %96.0
Health Hazard 1Warning