Hexamethylenetetramine, 99%
Hexamethylenetetramine, 99%
Hexamethylenetetramine, 99%
Hexamethylenetetramine, 99%
Thermo Scientific Chemicals
Hexamethylenetetramine, 99%
CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Plastic Bottle
Catalog number FSA120610025
Price (MYR)
620.00
BOT
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
620.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHexamethylenetetramine
CAS100-97-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Flammable solid.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.