1,1,1,3,3,3-Hexamethyldisilazane, 98%
1,1,1,3,3,3-Hexamethyldisilazane, 98%
1,1,1,3,3,3-Hexamethyldisilazane, 98%
1,1,1,3,3,3-Hexamethyldisilazane, 98%
Thermo Scientific Chemicals
1,1,1,3,3,3-Hexamethyldisilazane, 98%
CAS: 999-97-3 | C6H19NSi2 | 161.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
10 mL
100 mL
2.5 L
Packaging:
View all
Glass bottle
Catalog number FSA120585000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1,1,1,3,3,3-Hexamethyldisilazane
CAS999-97-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with