1-Heptene, 98%
1-Heptene, 98%
1-Heptene, 98%
Thermo Scientific Chemicals
1-Heptene, 98%
CAS: 592-76-7 | C7H14 | 98.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA120391000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Heptene
CAS592-76-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0