1-Heptanol, 98%
1-Heptanol, 98%
1-Heptanol, 98%
Thermo Scientific Chemicals
1-Heptanol, 98%
CAS: 111-70-6 | C7H16O | 116.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA120362500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Heptanol
Acidity0.5mg KOH/g max.
CAS111-70-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning