Guanine, 99+%
Guanine, 99+%
Guanine, 99+%
Thermo Scientific Chemicals
Guanine, 99+%
CAS: 73-40-5 | C5H5N5O | 151.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA120251000
Price (MYR)
519.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
519.00
EA
Add to cart
Specifications
Chemical Name or MaterialGuanine
CAS73-40-5
Absorbancelambda max. 248nm, pH 1, 1.35 to 1.41 (A250/A260), 0.82 to 0.86 (A280/A260), 0.47 to 0.51 (A290/A260)
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: