View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycylglycine
Melting Point262.0°C to 264.0°C
Absorbance(10% aq. soln.) (1 cm cell), 0.01 max. at 405nm
Name Note99+%
CAS556-50-3