Glycinamide hydrochloride, 98%
Glycinamide hydrochloride, 98%
Glycinamide hydrochloride, 98%
Glycinamide hydrochloride, 98%
Thermo Scientific Chemicals
Glycinamide hydrochloride, 98%
CAS: 1668-10-6 | C2H6N2O·HCl | 110.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA120061000
Price (MYR)
1,739.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,739.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGlycinamide hydrochloride
Name Note98%
CAS1668-10-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: