3-Fluorobenzaldehyde, 98+%
3-Fluorobenzaldehyde, 98+%
3-Fluorobenzaldehyde, 98+%
3-Fluorobenzaldehyde, 98+%
Thermo Scientific Chemicals
3-Fluorobenzaldehyde, 98+%
CAS: 456-48-4 | C7H5FO | 124.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Packaging:
Glass bottle
Catalog number FSA119330500
Price (MYR)
1,074.00
BOT
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,074.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Fluorobenzaldehyde
CAS456-48-4
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0