2-Ethyl-1,3-hexanediol, 99%, mixture of isomers
2-Ethyl-1,3-hexanediol, 99%, mixture of isomers
2-Ethyl-1,3-hexanediol, 99%, mixture of isomers
2-Ethyl-1,3-hexanediol, 99%, mixture of isomers
Thermo Scientific Chemicals
2-Ethyl-1,3-hexanediol, 99%, mixture of isomers
CAS: 94-96-2 | C8H18O2 | 146.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA118512500
Price (MYR)
275.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
275.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Ethyl-1, 3-hexanediol
Name Note99%
Acidity0.05mg KOH/g max.
CAS94-96-2
CAS Min %98.5