Ethyl 2-bromoisobutyrate, 98%
Ethyl 2-bromoisobutyrate, 98%
Ethyl 2-bromoisobutyrate, 98%
Ethyl 2-bromoisobutyrate, 98%
Thermo Scientific Chemicals
Ethyl 2-bromoisobutyrate, 98%
CAS: 600-00-0 | C6H11BrO2 | 195.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA118135000
Price (MYR)
1,215.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,215.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl 2-bromoisobutyrate
Name Note98%
CAS600-00-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause genetic defects if inhaled.