2(2-Ethoxyethoxy)ethanol, 98+%
2(2-Ethoxyethoxy)ethanol, 98+%
2(2-Ethoxyethoxy)ethanol, 98+%
2(2-Ethoxyethoxy)ethanol, 98+%
Thermo Scientific Chemicals
2(2-Ethoxyethoxy)ethanol, 98+%
CAS: 111-90-0 | C6H14O3 | 134.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA117890025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material2(2-Ethoxyethoxy)ethanol
CAS111-90-0
CAS Min %98.0
CAS Max %100.0
Health Hazard 2GHS H Statement: