meso-Erythritol, 99%
meso-Erythritol, 99%
meso-Erythritol, 99%
meso-Erythritol, 99%
Thermo Scientific Chemicals
meso-Erythritol, 99%
CAS: 149-32-6 | C4H10O4 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
View all
Plastic bottle
Catalog number FSA117821000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Materialmeso-Erythritol
Name Note99%
CAS149-32-6
Melting Point117.0°C to 121.0°C