Eicosanoic acid, 98%
Eicosanoic acid, 98%
Eicosanoic acid, 98%
Eicosanoic acid, 98%
Thermo Scientific Chemicals
Eicosanoic acid, 98%
CAS: 506-30-9 | C20H40O2 | 312.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA117720100
Price (MYR)
1,217.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,217.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEicosanoic acid
CAS506-30-9
Melting Point74.0°C to 77.0°C
ColorWhite
EINECS Number208-031-3