Dithiooxamide, 98%
Dithiooxamide, 98%
Dithiooxamide, 98%
Thermo Scientific Chemicals
Dithiooxamide, 98%
CAS: 79-40-3 | C2H4N2S2 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number FSA117551000
Price (MYR)
3,186.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
3,186.00
EA
Add to cart
Specifications
Chemical Name or MaterialDithiooxamide
CAS79-40-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Harmful if swallowed. Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation.
Health Hazard 3GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.