Dipropylamine, 99%
Dipropylamine, 99%
Dipropylamine, 99%
Thermo Scientific Chemicals
Dipropylamine, 99%
CAS: 142-84-7 | C6H15N | 101.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA117492500
Price (MYR)
207.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
207.00
EA
Add to cart
Specifications
Chemical Name or MaterialDipropylamine
Name Note99%
CAS142-84-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
May cause respiratory irritation.
Highly flammable liquid and vapor.