9,10-Diphenylanthracene, 98%
9,10-Diphenylanthracene, 98%
9,10-Diphenylanthracene, 98%
9,10-Diphenylanthracene, 98%
Thermo Scientific Chemicals
9,10-Diphenylanthracene, 98%
CAS: 1499-10-1 | C26H18 | 330.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA117200050
Price (MYR)
1,149.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,149.00
BOT
Add to cart
Specifications
Chemical Name or Material9, 10-Diphenylanthracene
Name Note99%
CAS1499-10-1
CAS Min %97.5
CAS Max %100.0