2,4-Dinitroaniline, 99%
2,4-Dinitroaniline, 99%
2,4-Dinitroaniline, 99%
2,4-Dinitroaniline, 99%
Thermo Scientific Chemicals
2,4-Dinitroaniline, 99%
CAS: 97-02-9 | C6H5N3O4 | 183.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA116935000
Price (MYR)
2,339.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,339.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 4-Dinitroaniline
Name Note99%
CAS97-02-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger