1,3-Dimethylurea, 98%
1,3-Dimethylurea, 98%
1,3-Dimethylurea, 98%
1,3-Dimethylurea, 98%
Thermo Scientific Chemicals
1,3-Dimethylurea, 98%
CAS: 96-31-1 | C3H8N2O | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA116911000
Price (MYR)
261.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
261.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 3-Dimethylurea
Name Note98%
CAS96-31-1
CAS Min %97.5
CAS Max %100.0