4,5-Dimethyl-o-phenylenediamine, 98%
4,5-Dimethyl-o-phenylenediamine, 98%
4,5-Dimethyl-o-phenylenediamine, 98%
4,5-Dimethyl-o-phenylenediamine, 98%
Thermo Scientific Chemicals
4,5-Dimethyl-o-phenylenediamine, 98%
CAS: 3171-45-7 | C8H12N2 | 136.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA116610250
Price (MYR)
2,066.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,066.00
EA
Add to cart
Specifications
Chemical Name or Material4, 5-Dimethyl-o-phenylenediamine
Melting Point127.2°C to 128.3°C
CAS3171-45-7
CAS Min %97.5
ColorBrown to White