2,5-Dimethylphenol, 99+%
2,5-Dimethylphenol, 99+%
2,5-Dimethylphenol, 99+%
2,5-Dimethylphenol, 99+%
Thermo Scientific Chemicals
2,5-Dimethylphenol, 99+%
CAS: 95-87-4 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA116540050
Price (MYR)
222.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
222.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 5-Dimethylphenol
Melting Point75°C to 77°C
Name Note99+%
CAS95-87-4
CAS Min %99.0