N,O-Dimethylhydroxylamine hydrochloride, 98%
N,O-Dimethylhydroxylamine hydrochloride, 98%
N,O-Dimethylhydroxylamine hydrochloride, 98%
N,O-Dimethylhydroxylamine hydrochloride, 98%
Thermo Scientific Chemicals
N,O-Dimethylhydroxylamine hydrochloride, 98%
CAS: 6638-79-5 | C2H8ClNO | 97.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA116340250
Price (MYR)
613.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
613.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, O-Dimethylhydroxylamine hydrochloride
Name Note98%
CAS6638-79-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
View more
Coupling reagent
RUO – Research Use Only