View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN, N-Dimethylethanolamine
CAS108-01-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0