2,6-Dimethylaniline, 99%
2,6-Dimethylaniline, 99%
2,6-Dimethylaniline, 99%
2,6-Dimethylaniline, 99%
Thermo Scientific Chemicals
2,6-Dimethylaniline, 99%
CAS: 87-62-7 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA115945000
Price (MYR)
844.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
844.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 6-Dimethylaniline
Melting Point10°C to 12°C
Name Note99%
CAS87-62-7
CAS Min %98.5