N,N-Dimethylaniline, 99%
N,N-Dimethylaniline, 99%
N,N-Dimethylaniline, 99%
N,N-Dimethylaniline, 99%
Thermo Scientific Chemicals
N,N-Dimethylaniline, 99%
CAS: 121-69-7 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA115920025
Price (MYR)
554.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
554.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethylaniline
CAS121-69-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0