2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers
2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers
2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers
2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers
Thermo Scientific Chemicals
2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers
CAS: 696-59-3 | C6H12O3 | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA115671000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 5-Dimethoxytetrahydrofuran
Melting Point-45°C
Name Note99%
CAS696-59-3
CAS Min %98.5