Diethyl phthalate, 99%
Diethyl phthalate, 99%
Diethyl phthalate, 99%
Diethyl phthalate, 99%
Thermo Scientific Chemicals
Diethyl phthalate, 99%
CAS: 84-66-2 | C12H14O4 | 222.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA114520250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethyl phthalate
Name Note99%
CAS84-66-2
Density1.1180g/mL
RTECS NumberTI1050000