Diethyl acetylsuccinate, 99%
Diethyl acetylsuccinate, 99%
Diethyl acetylsuccinate, 99%
Diethyl acetylsuccinate, 99%
Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%
CAS: 1115-30-6 | C10H16O5 | 216.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA113951000
Price (MYR)
1,229.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,229.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiethyl acetylsuccinate
CAS1115-30-6
Melting Point-8°C
ColorYellow
Density1.0800g/mL