Diethanolamine, 99%
Diethanolamine, 99%
Diethanolamine, 99%
Thermo Scientific Chemicals
Diethanolamine, 99%
CAS: 111-42-2 | C4H11NO2 | 105.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Glass Bottle
Catalog number FSA113920025
Price (MYR)
472.00
BOT
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
472.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiethanolamine
CAS111-42-2
Health Hazard 1Danger
Health Hazard 2GHS P Statement
Causes serious eye damage.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Health Hazard 3Emergency Overview
Harmful if swallowed. Irritating to eyes and skin. Risk of serious damage to eyes. May cause irritation of respiratory tract. Corrosive to metals.
NFPA&C