N,N'-Dicyclohexylcarbodiimide, 99%
N,N'-Dicyclohexylcarbodiimide, 99%
N,N'-Dicyclohexylcarbodiimide, 99%
N,N'-Dicyclohexylcarbodiimide, 99%
Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 99%
N,N-Dicyclohexylcarbodiimide, 99%, C13H22N2, CAS Number-538-75-0, carbodiimide, dicyclohexyl, bis cyclohexyl carbodiimide, dcc, 1,3-dicyclohexylcarbodiimide, carbodicyclohexylimide, n,n'-dicyclohexylcarbodiimide, n,n-dicyclohexylcarbodiimide, dcci, dicyclohexylcarbodiimide, dccd, 1kg | CAS: 538-75-0 | C13H22N2 | 206.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
5 kg
25 kg
Packaging:
View all
Glass bottle
Catalog number FSA113902500
Price (MYR)
786.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
786.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN,N′-Dicyclohexylcarbodiimide
CAS538-75-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Coupling reagent
RUO – Research Use Only