Dicyandiamide, 99.5%
Dicyandiamide, 99.5%
Dicyandiamide, 99.5%
Dicyandiamide, 99.5%
Thermo Scientific Chemicals
Dicyandiamide, 99.5%
CAS: 461-58-5 | C2H4N4 | 84.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA113810250
Price (MYR)
264.00
EA
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
264.00
EA
Add to cart
Specifications
Chemical Name or MaterialDicyandiamide
CAS461-58-5
Health Hazard 1
Melting Point208.0°C to 211.0°C