Dichloromethane, 99.8+%, for analysis, stabilized with amylene
Dichloromethane, 99.8+%, for analysis, stabilized with amylene
Dichloromethane, 99.8+%, for analysis, stabilized with amylene
Dichloromethane, 99.8+%, for analysis, stabilized with amylene
Thermo Scientific Chemicals
Dichloromethane, 99.8+%, for analysis, stabilized with amylene
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA113460010
Price (MYR)
483.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
483.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDichloromethane
Melting Point-97.0°C
CAS75-09-2
Health Hazard 1Warning
Health Hazard 2GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure.
View more
For analysis
RUO – Research Use Only