Dichlorodimethylsilane, 99+%
Dichlorodimethylsilane, 99+%
Dichlorodimethylsilane, 99+%
Dichlorodimethylsilane, 99+%
Thermo Scientific Chemicals
Dichlorodimethylsilane, 99+%
CAS: 75-78-5 | C2H6Cl2Si | 129.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA113312500
Price (MYR)
1,991.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,991.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDichlorodimethylsilane
CAS75-78-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p