View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2,6-Di-tert-butyl-4-methylphenol
CAS128-37-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0