alpha,alpha'-Dibromo-o-xylene, 96%
alpha,alpha'-Dibromo-o-xylene, 96%
alpha,alpha'-Dibromo-o-xylene, 96%
alpha,alpha'-Dibromo-o-xylene, 96%
Thermo Scientific Chemicals
alpha,alpha'-Dibromo-o-xylene, 96%
CAS: 91-13-4 | C8H8Br2 | 263.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA112930250
Price (MYR)
588.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
588.00
BOT
Add to cart
Specifications
Chemical Name or Materialα, α'-Dibromo-o-xylene
Name Note96%
CAS91-13-4
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger